Learn More
Chemglass Life Sciences Filter Adapter, Neoprene, Size 7
SDP
Supplier:  Chemglass Life Sciences CG140116
Encompass_Preferred
Filter Adapter, Neoprene, Size 7. Height: 45mm ID Top/Bottom (mm): 75 x 46 OD Top/Bottom (mm): 86 x 57 Wall thickness: 5.5mm Neoprene filter adapters are designed to fit between a filtering funnel and a flask. For use with Buchner funnels, Gooch crucibles, filter tubes, etc. Can be used individually or nested with adjacent sizes. Eliminates the boring of rubber stoppers and the difficulties of inserting and removing glass stems in stoppers. Neoprene withstands autoclaving. Available as a set or individually.
Catalog No. CG140116