Learn More
Wilmad Labglass FILTER APP 47MM 1000ML/4L
Supplier:  Wilmad Labglass BP1825004
Encompass_Preferred
Filter Apparatus, All-Glass. Filter Size: 47mm; Funnel Capacity: 1000mL; Flask Capacity: 4000mL; Joint Size: [ST] 40/35. All Glass filter apparatus w/ standard taper joint connecting the membrane filter support base to the filter flask
Catalog No. 50-805-282
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov