Learn More
Wilmad Labglass FILTER APP 47MM 500ML/2L
Supplier:  Wilmad Labglass BP1825002
Encompass_Preferred
Filter Apparatus, All-Glass. Filter Size: 47mm; Funnel Capacity: 500mL; Flask Capacity: 2000mL; Joint Size: [ST] 40/35. All Glass filter apparatus w/ standard taper joint connecting the membrane filter support base to the filter flask
Catalog No. 50-805-333