Learn More
Pall Supor EKV membrane in Mini Kleenpak™ capsules, 0.2 µm removal rating, 200 cm² EFA, ¼-½ in. hose barb connections, pre-sterilized by gamma irradiation, box of 3
Supplier:  Pall KA02EKVP2S
Encompass
Capsule Filtration of Buffers, Tissue Culture Media, and Other Biological Fluids
Catalog No. NC9669902