Learn More
STEMCELL Technologies Filter Cards and Spacers, Size: 48 Units
SDP
Supplier:  STEMCELL Technologies 04911
Encompass
Use Filter Cards and Spacers for the dehydration of collagen-based gels following culture.Available either individually or as part of the Double Chamber Slide Kit (/products/double-chamber-slide-kit.html), the Filter Cards and Spacers are intended to be used with MegaCult™-C Double Chamber Slides (/products/megacult-c-double-chamber-slides.html) for performing colony assays in collagen gels for the detection of hematopoietic progenitor cells.
Catalog No. 50-197-4332