Learn More
Applied Separations FILTER COLUMN 35ML 20PE 100/PK
SDP
Supplier:  Applied Separations 2435
Encompass_Preferred
Cartridge, SPE; Applied Separations; w/one hydrophobic 20m PE frit; Polypropylene; Size: 35mL; Quantity: 100/pk.
Catalog No. 50-900-094