Learn More
Applied Separations FILTER COLUMN 3ML 20PE 100/PK
SDP
Supplier:  Applied Separations 2413
Encompass_Preferred
Cartridge, SPE; Applied Separations; w/one hydrophobic 20m PE frit; Polypropylene; Size: 3mL; Quantity: 100/pk.
Catalog No. 50-900-090