Learn More
Chemglass Life Sciences 150mL Filter Funnel, Jacketed, Medium Frit
SDP
Supplier:  Chemglass Life Sciences CG140311
Encompass_Preferred
150mL Filter Funnel, Jacketed, Medium Frit. Similar to CG-1402 but with a water jacket enclosing the funnel bowl and upper portion of the stem. Jacket permits heating or cooling of the contents of the funnel during filtration. Stem length listed is the length below the jacket. Jacketed filter funnels are available in sizes other than those listed. Please call for details.
Catalog No. CG140311