Learn More
Chemglass Life Sciences Filter Funnel, Hirsh, 75mm Diameter Funnel Top, 30mm Fine Frit
SDP
Supplier:  Chemglass Life Sciences CG140808
Encompass_Preferred
Filter Funnel, Hirsh, 75mm Diameter Funnel Top, 30mm Fine Frit. Used in applications where it is necessary to wash and re-dissolve the precipitate with chemicals that would attack standard filter paper. The 60º angle on the funnel permits removal of precipitates.
Catalog No. CG140808