Learn More
Ace Glass, Inc. Filter funnel, Instatherm, 500mL, 40v/6amp max
SDP
Supplier:  Ace Glass, Inc. 370401
Encompass
500mL Instatherm filter funnel, for use with the Instatherm filtration apparatus assembly. The Instatherm filter funnel keeps the viscous material, such as polymers, in the liquid state.
Catalog No. 50-971-350