Learn More
IDEX Health & Science Upchurch FILTER GENERAL SOLVENT INLET
Supplier:  IDEX Health & Science Upchurch A302A
Encompass_Preferred
Idex Inlet Solvent Filters, Includes: 1 Filter (Cups and end fittings sold separately), Pore Size: 10 um, Flow Rate: 40 mL/min, Stainless Steel, Thread Size: 0.25 to 28 Flat bottom
Catalog No. NC1920932