Learn More
Hach Company Hach Glass Fiber Filters, 110mm, 100/pk
Supplier:  Hach Company 2511211
Encompass
Hach glass microfiber filters are designed to exceed ASTM and Standard Methods protocols including 2540 C, 2540 D, and 2540 E. Our filters have a smooth surface and are binder-free with unique high temperature resistance. The 1.0 µm retention offers fast speed as well as a very high loading capacity. Hach glass filters are designed to maintain a stable weight before and after treatment in excess of 550 °C. This is an important attribute for customers performing Fixed and Volatile Solids analysis of water and wastewater.
Catalog No. 50-199-6128