Learn More
Cole-Parmer Thermo Scientific 760209 Incubator Replacement HEPA Filter, 4/pk
Supplier:  Cole-Parmer 3932034
Encompass_Preferred
Heavy-duty wire shelves are available in stationary and pull-out options. Design assures air circulation to maintain temperature uniformity. Shelves can be used in 6.5 and 19.5 cu ft versions.
Catalog No. NC0769936