Learn More
Microsolv Technology Corporation Filter, Hplc Mobile Phase Inle
Supplier:  Microsolv Technology Corporation 5884601
Encompass_Preferred
Mobile Phase Filter, 10um, Last Drop, Stainless Steel, Tripod Tubing Connector. MicroSolv Brand. 1/EA.
Catalog No. NC1909605