Learn More
Microsolv Technology Corporation Filter, Hplc Mobile Phase Inlt
Supplier:  Microsolv Technology Corporation 5884607
Encompass_Preferred
Mobile Phase Filter, 5.0um, Last Drop, PTFE, Tripod Tubing Connector. MicroSolv Brand. 1/EA.
Catalog No. NC1909607