Learn More
Optimize Technologies Filter Insert Assembly, 2µm, Waters 2690/2695, 2790/2795, 600
SDP
Supplier:  Optimize Technologies 390403524
Encompass_Preferred
Most instrument manufacturers will tell you quite emphatically that for optimal instrument performance, you should only use components that match their performance specifications. We could not agree more. Every replacement component we offer is designed to meet or exceed original equipment specifications - guaranteed. Key features for this item include an In-line Filer Insert Assembly with 2µm porosity. It is designed to work with Waters 2690/2695, 2790-2795 and 600 pump models-equivalent to OEM part number WAT088084.
Catalog No. 50-200-6697