Learn More
Andwin Scientific FILTER PAPER 4.25 BLACK 100/PK
SDP
Supplier:  Andwin Scientific 28342012
Encompass_Preferred
The Black Qualitative Filter Paper features a smooth black finish that is ideal for use in procedures that need visual examination of particles. This product has a diameter of 4.25 centimeters and is sold in a pack of 100.
Catalog No. NC0615192