Learn More
Bio Rad Laboratories FILTER PAPER 7.5 X 10CM
Supplier:  Bio Rad Laboratories 1703965
Encompass
Blot Absorbent Filter Paper, 60 sheets, 7.5 x 10cm, Extra thick, Fits Mini-PROTEAN 3, Recommended for all blotting applications using the Trans-Blot SD semidry transfer cell or Trans-Blot
Catalog No. NC9418388