Learn More
Hach Company QL Filters, Pore Size 8 - 12 µm, Diameter 90 mm, Circular, 100/pk, Qualitative
Supplier:  Hach Company 50655
Encompass
QL Filters are recommended for use in various environmental and wet chemistry reference methods. Extremely pure cellulose filters of nearly 100% alpha content are specially processed to yield negligible extractable levels in the determination and identification of unknown sample materials.
Catalog No. 50-199-4339