Learn More
Hach Company SA Filter, Cone, Diameter 110 mm, Soil Analysis Grade, 500/pk
Supplier:  Hach Company 2263356
Encompass
Designed for sandy soils, SA filters are conically folded and have been optimized for use with the common extraction solutions used in soil nutrient analysis (Mehlich 1, Mehlich 3, Morgan, Modified Morgan, Bray, and Olsen). SA Filters provide the best combination of retention rating, filtration speed and purity requirements for soil nutrient testing (P, K, Ca, Mg).
Catalog No. 50-199-5663