Learn More
Thomson Instrument Company Thomson 96-Well Filter Plate, 2mL, Square Well, Long Drip | 25µm Polypropylene Filter
SDP
Supplier:  Thomson Instrument Company 931919
Encompass_Preferred
Thomson Filter Plates (24- and 96-well) enable fast, parallel plate-to-plate processing for biological and analytical workflows. Within our portfolio: Rapid Clear 0.2 µm depth filters for cell-culture clarification by centrifuge, PTFE/PVDF membranes for final filtration, and 25 µm polypropylene plates for resin/adsorption purification. Pair with Thomson Collection Plates and seals for clean transfers and high-throughput screening.
• 96-well format with 2 mL working volume per well.
• Square Well, Long Drip well design.
• 25 µm polypropylene filter membrane.
• ANSI/SLAS compliant — automation compatible.
• Use with collection plate 931130.
• Non-sterile.
• Made in the USA
Catalog No. NC1254587