Learn More
Hach Company PTFE Support Plate for Filtering Funnel Assembly
Supplier:  Hach Company 2164200
Encompass
Optional PTFE support plate for acid and neutral detergent fiber tests. For use with the Filter Funnel Assembly, 2164100.
Catalog No. 50-199-5488