Learn More
IDEX Health & Science Upchurch FILTER PRECOLUMN UPTIGHT
Supplier:  IDEX Health & Science Upchurch A315
Encompass_Preferred
Idex Precolumn Inline Filters, Pore Size: 2um, Housing Material: Stainless Steel, Mass: Swept Volume: 0.66uL, Length: 0.94 in.
Catalog No. NC1920937