Learn More
Cole-Parmer Minntech 279-30-105 Hollow Fiber Filter Capsule, 1-5.7L, 0.2um, 3/8"NPTM; 6/Pk
Supplier:  Cole-Parmer 2988038
Encompass_Preferred
These new hollow fiber filters offer superior performance in a compact size. Filters have up to six times more surface area compared to other pleated membrane filters, providing greater capacity and longer filter life. The unique membrane structure helps remove bacteria, high-level endotoxins, pyrogens, and spores ideal for a variety of water purification applications. Maximum differential pressure is 30 psi (2 bar). Capsules and cartridges provided nonsterile.
Catalog No. NC1676667