Learn More
Waters Corp Solvent Filter, PTFE, 47 mm, 0.45 µm, Non polar, 100/pk
Supplier:  Waters Corp WAT200534
Encompass_Preferred
This PTFE membrane is a good choice for use with aggresive organic solvents
Catalog No. 50-786-077