Learn More
Andwin Scientific FILTER REPLACE SET 0.45UM PK5
SDP
Supplier:  Andwin Scientific JENC266003
Encompass_Preferred
The Membrane Filters are ideal for use with the Safetypette and Powerpette pipet controllers. This product has a pore size of 0.45 um. The filters protect the controllers from fluid over-aspiration and thus lessen the chance of cross-contamination. This is sold in the pack of 5.
Catalog No. 50-104-7967