Learn More
Microsolv Technology Corporation Filter Sf .22 Ptfe 13Mm Blue 1
Supplier:  Microsolv Technology Corporation 58022P13C
Encompass_Preferred
Syringe Filters, 13mm, PTFE, 0.22um Pore Size. Blue Polypropylene, AQ Brand. 100/PK.
Catalog No. 50-014-52581