Learn More
Microsolv Technology Corporation Filter Sf .22 Ptfe 4Mm Blue 10
Supplier:  Microsolv Technology Corporation 58022P04C
Encompass_Preferred
Syringe Filters, 4mm, PTFE, 0.22um Pore Size. Blue Polypropylene, AQ Brand. 100/PK.
Catalog No. 50-014-52579