Learn More
Microsolv Technology Corporation Filter Sf.45 Nylon 13Mm Grn 10
Supplier:  Microsolv Technology Corporation 58045N13K
Encompass_Preferred
Syringe Filters, 13mm, Nylon, 0.45um Pore Size. Green Polypropylene, AQ Brand. 1000/CS.
Catalog No. 50-014-52600