Learn More
Microsolv Technology Corporation Filter Sf .45 Ptfe 25Mm Yel 1K
Supplier:  Microsolv Technology Corporation 58045P25K
Encompass_Preferred
Syringe Filters, 25mm, PTFE, 0.45um Pore Size. Yellow Polypropylene, AQ Brand. 1000/CS.
Catalog No. 50-014-52612