Learn More
Microsolv Technology Corporation Filter Sf .45 Pvdf 25Mm Red 1K
Supplier:  Microsolv Technology Corporation 58045V25K
Encompass_Preferred
Syringe Filters, 25mm, PVDF, 0.45um Pore Size. Red Polypropylene, AQ Brand. 1000/CS.
Catalog No. 50-014-52626