Learn More
TWD Scientific LLC BioX 500mL Vacuum Filter System. Case of 12
SDP
Supplier:  TWD Scientific LLC BX154500
Encompass_Preferred
BioX 500ml filter system, pes filter material, 0.22mm pore size, 75mm membrane diameter, sterile, individually wrapped. 12/case, price/case
Catalog No. 50-227-4285