Learn More
Chemglass Life Sciences Filter Tube, 24/40 Inner Joints, 40mm OD Medium Frit, 2mm Glass Stpks, Airfree, Schlenk
SDP
Supplier:  Chemglass Life Sciences AF054608
Encompass_Preferred
Filter Tube, 24/40 Inner Joints, 40mm OD Medium Frit, 2mm Glass Stpks, Airfree, Schlenk. Similar to AF-0544, but with the addition of two side arms, 2mm high vacuum glass stopcocks having a Stainless Steel constant pressure retaining devices.
Catalog No. AF054608