Learn More
Chemglass Life Sciences Filter Tube, Allihn, Fine Frit
SDP
Supplier:  Chemglass Life Sciences CG141003
Encompass_Preferred
Filter Tube, Allihn, Fine Frit. Allihn type filter tube with a 30mm O.D. disc. Height above the disc is 100mm, lower stem is 8mm O.D. x 55mm in length.
Catalog No. CG141003