Learn More
Thomson Instrument Company Standard|Filter Vial,PVDF 0.2µm, Pre-Slit Septum, Red Cap
SDP
Supplier:  Thomson Instrument Company 35531200
Encompass_Preferred
Standard|Filter Vials are designed to speed up sample prep and minimize transfer steps. Available in a variety of membranes and pore sizes. Simply add sample and diluent, press, and it is ready for analysis. This process minimizes sample loss. Pre-slit caps ensure a no-hassle, clean withdraw.Recommended for samples containing less than 10% solid particulates.
•Simply add sample, diluent, and press
•Color-coded caps allow easy identification of 0.2 or 0.45 μm membranes in PVDF, PTFE, PES, or Nylon
•Fits most standard 12×32 mm autosamplersThe Standard|Filter Vials are made from polypropylene and come in a variety of membranes and pore sizes.
• Made in the USA
Catalog No. 50-751-6347