Learn More
Chemglass Life Sciences 500mL Filtering Flask, Heavy Wall, 24/40 Outer Joint, Hose Connection
SDP
Supplier:  Chemglass Life Sciences CG155004
Encompass_Preferred
500mL Filtering Flask, Heavy Wall, 24/40 Outer Joint, Hose Connection. HEAVY WALL Erlenmeyer shaped filtering flask with standard taper outer joint and serrated hose connection for attachment to vacuum source. For stoppers see CG-3000.
Catalog No. CG155004