Learn More
Hach Company Sterile Millipore S-Pak Type HA Membrane Filters, 47 mm, 0.45 µm, gridded, pk/600
Supplier:  Hach Company 2936500
Encompass
For membrane filtration in microbiological analysis of aqueous samples. "HA" signifies certified compliance with Standard Methods criteria with pore size of 0.45 um. White, cellulose and mixed-ester composition. S-Pak (filter only) is sterile. Diameter: 4
Catalog No. 50-199-6842