Learn More
Microsolv Technology Corporation Filters, Membrane, 47mm, 0.45um, PVDF. Round, 100/PK.
Supplier:  Microsolv Technology Corporation 58445VD47
Encompass_Preferred
These high flow, non media migrating, Polyvinylidene Fluoride (PVDF) Advanced Quality (AQ) Brand, Filter Discs can be used with the Mobile Phase Filter Degasser Assembly or any device when membrane filtration is desired.Hydrophilic, they are low protein binding and can be autoclaved at 121⁰C for 30 minutes or EO sterilization without damage to the integrity or performance of the filter. Diameter: 47mm Max Operating Temperature: 80⁰C Bubble Point: 30psi (2.10 kg/cm2) Water Flow Rate: 30ml/min/cm2 @ 10 psi, 27⁰C Thickness: 135um +- 15um Extractables with WFI (water): Within USP Limits Oxidizable Substances: Passes USP Test Pore Size: 0.45 um
Catalog No. 50-227-1187