Learn More
National Diagnostics FILTON-X 1 GL
SDP
Catalog No. 5089990173
Encompass
Filtron-X, Ready-to-use liquid scintillation solution which completely dissolves Millipore-type filters, 1 GL
Catalog No. 50-899-90173 Supplier National Diagnostics Supplier No. LS2011GLL
Filtron-X, Ready-to-use liquid scintillation solution which completely dissolves Millipore-type filters, 1 GL