Learn More
Andwin Scientific FILTR VERSAPOR 3UM 25MM PK/100
SDP
Catalog No. NC0648730
Encompass_Preferred
The 3um/25mm Versapor 3000 Membrane Disc Filter is ideal for prefiltration of difficult to filter solutions and serum. It can reduce clogging of final filters.
Catalog No. NC0648730 Supplier Andwin Scientific Supplier No. 66386
The 3um/25mm Versapor 3000 Membrane Disc Filter is ideal for prefiltration of difficult to filter solutions and serum. It can reduce clogging of final filters.