Learn More
Sterlitech Corporation Filtration Adaptor for VF12, Fits GL45 thread media bottle (197000-65)
SDP
Supplier:  Sterlitech Corporation 19700065
Encompass_Preferred
This filtration adaptor design enables VF12 glass filter funnel connection with GL45 storage bottle. Constructed with autoclavable polypropylene for easy cleaning and sterilization.
Catalog No. NC1816049