Learn More
Azzota Corp 2 Liter Filtration Apparatus, Mobile Phase Filtration Apparatus Includes 2000Ml Flask With 500Ml Funnel , 47mm Diameter Membrane, Glass Connection With Clamp
Supplier:  Azzota Corp FA2000
Encompass_Preferred
2 Liter Filtration Apparatus, Mobile Phase Filtration Apparatus includes 2000ml flask with 500ml funnel , 47mm Diameter membrane, Glass connection with Clamp
Catalog No. 50-753-2963