Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 12mL Cartridge with 1/16 20m Frit

Catalog No. 502398704 Shop All Phenomenex Products
Encompass_Preferred

Filtration Column 12mL FC012

Catalog No. 50-239-8704 Supplier Phenomenex Inc Supplier No. FC012
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Filtration Column 12mL FC012

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.