Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 25mL Cartridge with 2 x 1/16 20m Frit

Catalog No. 502398532 Shop All Phenomenex Products
Encompass_Preferred

Filtration Column 25mL FC025

Catalog No. 50-239-8532 Supplier Phenomenex Inc Supplier No. FC025
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Filtration Column 25mL FC025

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.