Learn More
Cole-Parmer Kinesis KX Filtration System, 300 mL / 1 L
Catalog No. 501961433 Shop All Cole-Parmer Products
Encompass_Preferred
Use this all-glass filtration system for your work with particulate, microbiological, and HPLC to minimize contact between material and sample. 47 mm glass membrane funnel mounts on a flask reservoir with a ground glass joint. The glass membrane funnels drip tube is positioned below the barb to stop any sample aspiration in to the vacuum hose. Clean the all-glass system with organic and aqueous solvents.
Catalog No. 50-196-1433 Supplier Cole-Parmer Supplier No. 1291812
Use this all-glass filtration system for your work with particulate, microbiological, and HPLC to minimize contact between material and sample. 47 mm glass membrane funnel mounts on a flask reservoir with a ground glass joint. The glass membrane funnels drip tube is positioned below the barb to stop any sample aspiration in to the vacuum hose. Clean the all-glass system with organic and aqueous solvents.