Learn More
Cole-Parmer Kinesis KX Filtration System, 500 mL / 2 L
Supplier:  Cole-Parmer 1291814
Encompass_Preferred
Use our all-glass filtration systems for your work with particulate, microbiological, and HPLC to minimize contact between material and sample. 47 mm glass membrane funnel mounts on a flask reservoir with a ground glass joint. The glass membrane funnels drip tube is positioned below the " barb to stop any sample aspiration in to the vacuum hose. Clean the all-glass system with organic and aqueous solvents.
Catalog No. 50-196-1434