Learn More
Nacalai USA Inc Centrifugal filtration plate for 96-well, 100 µm, 50 plates
Catalog No. NC3787209
Encompass
For folow cytometer users, this is a mesh-equipped microplate in which specimens are dispensed by multichannel pipette, automatic dispenser, etc. with stacked on another microplate, then filtrated at one time by centrifuge. This is convenient for those who want to operate more easily and with less time.Mesh size: 100 µmMaterial: Polystylene (Plate), Polyester fiber (Mesh), Silicone (Adhesive)Compatible with 96 well plate
Catalog No. NC3787209 Supplier Nacalai USA Inc Supplier No. RCSC08003
For folow cytometer users, this is a mesh-equipped microplate in which specimens are dispensed by multichannel pipette, automatic dispenser, etc. with stacked on another microplate, then filtrated at one time by centrifuge. This is convenient for those who want to operate more easily and with less time.Mesh size: 100 µmMaterial: Polystylene (Plate), Polyester fiber (Mesh), Silicone (Adhesive)Compatible with 96 well plate