Learn More
TARGETMOL CHEMICALS INC FIN56 25MG
SDP
Catalog No. 502579978
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FIN56 is a specific inducer of ferroptosis. purity: 99%
Catalog No. 50-257-9978 Supplier TARGETMOL CHEMICALS INC Supplier No. T406625MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FIN56 is a specific inducer of ferroptosis. purity: 99%