Learn More
Electron Microscopy Sciences Final B Polishing Cloth PSA with 2.5" center hole, 8"
SDP
Catalog No. 50997228
Encompass_Preferred
This cloth has been especially designed for long life. Made from a rayon flock on a cotton canvas backing. It is recommended for all final polishing. Available with Pressure Sensitive Adhesive (PSA).
Type: 2.5" centered hole
Diameter: 8"
Catalog No. 50-997-228 Supplier Electron Microscopy Sciences Supplier No. 5033415
This cloth has been especially designed for long life. Made from a rayon flock on a cotton canvas backing. It is recommended for all final polishing. Available with Pressure Sensitive Adhesive (PSA).
Type: 2.5" centered hole
Diameter: 8"