Learn More
Electron Microscopy Sciences Final B Polishing Cloth PSA backing and no hole, 8"
SDP
Catalog No. 50997227
Encompass_Preferred
This cloth has been especially designed for long life. Made from a rayon flock on a cotton canvas backing. It is recommended for all final polishing. Available with Pressure Sensitive Adhesive (PSA).
Type: no hole
Diameter: 8"
Catalog No. 50-997-227 Supplier Electron Microscopy Sciences Supplier No. 5033405
This cloth has been especially designed for long life. Made from a rayon flock on a cotton canvas backing. It is recommended for all final polishing. Available with Pressure Sensitive Adhesive (PSA).
Type: no hole
Diameter: 8"